1-Benzoyl-pyrrolidin-2-one 95%

Lieferant: Apollo Scientific

OR930749-1G
APOSOR930749-1GEA 0 EUR
APOSOR930749-1G
1-Benzoyl-pyrrolidin-2-one 95%
1-Benzoyl-pyrrolidin-2-one
Formel: C₁₁H₁₁NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00030544
CAS-Nummer: 2399-66-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR