3-Chloro-5-(hydroxymethyl)benzonitrile 95%
Lieferant: Apollo Scientific
OR930750-1G
APOSOR930750-1GEA
94.4
EUR
APOSOR930750-1G
3-Chloro-5-(hydroxymethyl)benzonitrile 95%
3-Chloro-5-(hydroxymethyl)benzonitrile
Formel:
C₈H₆ClNO Molecular Weight: 167,592 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18392447 CAS-Nummer: 1021871-35-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...