3-(Methylthio)benzaldehyd 95%

Lieferant: Apollo Scientific

OR930751-1G OR930751-250MG
APOSOR930751-1GEA 159 EUR
APOSOR930751-1G APOSOR930751-250MG
3-(Methylthio)benzaldehyd 95%
3-(Methylthio)benzaldehyde
Formel: C₈H₈OS
Lagertemperatur: Raumtemperatur
MDL: MFCD09864229
CAS-Nummer: 73771-35-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR