4-Butylcyclohexanamin (cis/trans-Gemisch) 95%

Lieferant: Apollo Scientific

Synonyme: 1-Amino-4-butylcyclohexane

OR930768-5G
APOSOR930768-5GEA 464 EUR
APOSOR930768-5G
4-Butylcyclohexanamin (cis/trans-Gemisch) 95%
4-Butylcyclohexylamine (cis- and trans- mixture)
Formel: C₁₀H₂₁N
Molecular Weight: 155,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09261273
CAS-Nummer: 37942-69-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR