Propyl cyanoacetate 95%

Lieferant: Apollo Scientific

Synonyme: Propyl 2-cyanoacetate, Cyanoacetic Acid Propyl Ester

OR930774-100G
APOSOR930774-100GEA 68.3 EUR
APOSOR930774-100G
Propyl cyanoacetate 95%
Propyl cyanoacetate
Formel: C₆H₉NO₂
Molecular Weight: 127.14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00015784
CAS-Nummer: 14447-15-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR