3-(1H-Indol-1-yl)propanoic acid 95%

Lieferant: Apollo Scientific

OR930793-1G
APOSOR930793-1GEA 155 EUR
APOSOR930793-1G
3-(1H-Indol-1-yl)propanoic acid 95%
3-(1H-Indol-1-yl)propanoic acid
Formel: C₁₁H₁₁NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00022896
CAS-Nummer: 6639-06-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR