(4-Chloro-pyrimidin-2-yl)-cyclopropyl-amine 95%

Lieferant: Apollo Scientific

OR930797-1G
APOSOR930797-1GEA 0 EUR
APOSOR930797-1G
(4-Chloro-pyrimidin-2-yl)-cyclopropyl-amine 95%
(4-Chloro-pyrimidin-2-yl)-cyclopropyl-amine
Formel: C₇H₈ClN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11046853
CAS-Nummer: 1044771-76-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR