(1-Methyl-5-thien-2-yl-1H-pyrazol-3-yl)methanol 97%
Lieferant: Apollo Scientific
OR9308-1G
APOSOR9308-1GEA
371
EUR
APOSOR9308-1G
(1-Methyl-5-thien-2-yl-1H-pyrazol-3-yl)methanol 97%
(1-Methyl-5-thien-2-yl-1H-pyrazol-3-yl)methanol
Formel:
C₉H₁₀N₂OS Molecular Weight: 194,254 g/mol Schmelzpunkt: 56…60 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08435882 CAS-Nummer: 879896-47-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...