Allyl-dimethylcarbamodithioat 95%
Lieferant: Apollo Scientific
Synonyme:
Dimethyldithiocarbamic Acid Allyl Ester
OR930814-5G
APOSOR930814-5GEA
487
EUR
APOSOR930814-5G
Allyl-dimethylcarbamodithioat 95%
Allyl-dimethylcarbamodithioat
Formel:
C₆H₁₁NS₂ Molecular Weight: 161,28 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00059057 CAS-Nummer: 20821-66-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...