2-(1H-1,2,4-Triazol-3-yl)pyridin 95%
Lieferant: Apollo Scientific
Synonyme:
3-(2-Pyridyl)-1,2,4-triazole
OR930824-5G
APOSOR930824-5GEA
105
EUR
APOSOR930824-5G
2-(1H-1,2,4-Triazol-3-yl)pyridin 95%
2-(1H-1,2,4-Triazol-3-yl)pyridine
Formel:
C₇H₆N₄ Molecular Weight: 146.15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13188589 CAS-Nummer: 23195-62-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...