(1-Methyl-3-thien-2-yl-1H-pyrazol-5-yl)methanol 97%

Lieferant: Apollo Scientific

OR9309-1G
APOSOR9309-1GEA 371 EUR
APOSOR9309-1G
(1-Methyl-3-thien-2-yl-1H-pyrazol-5-yl)methanol 97%
(1-Methyl-3-thien-2-yl-1H-pyrazol-5-yl)methanol
Formel: C₉H₁₀N₂OS
Molecular Weight: 194,254 g/mol
Schmelzpunkt: 95…97 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08435885
CAS-Nummer: 879896-49-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR