2-Methyl-1-(p-tolyl)-2-propanol 95%

Lieferant: Apollo Scientific

Synonyme: alpha,alpha,4-Trimethylphenethyl Alcohol, 2-Methyl-1-(p-tolyl)propan-2-ol, 1,1-Dimethyl-2-(p-tolyl)ethanol

OR930931-5G
APOSOR930931-5GEA 201 EUR
APOSOR930931-5G
2-Methyl-1-(p-tolyl)-2-propanol 95%
2-Methyl-1-(p-tolyl)-2-propanol
Formel: C₁₁H₁₆O
Molecular Weight: 164.25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00029997
CAS-Nummer: 20834-59-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR