Isochinolin-3-carbonitril 95%

Lieferant: Apollo Scientific

Synonyme: 3-Isoquinolinecarbonitrile

OR930950-5G
APOSOR930950-5GEA 226 EUR
APOSOR930950-5G
Isochinolin-3-carbonitril 95%
Isochinolin-3-carbonitril
Formel: C₁₀H₆N₂
Molecular Weight: 154.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00075136
CAS-Nummer: 26947-41-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR