5-Methyl-2,3-dihydro-1H-inden-1-amine 95%

Lieferant: Apollo Scientific

OR930980-250MG
APOSOR930980-250MGEA 183 EUR
APOSOR930980-250MG
5-Methyl-2,3-dihydro-1H-inden-1-amine 95%
5-Methyl-2,3-dihydro-1H-inden-1-amine
Formel: C₁₀H₁₃N
Molecular Weight: 147,217 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07373907
CAS-Nummer: 168902-79-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR