4-(Boc-amino)pyridazine 95%
Lieferant: Apollo Scientific
OR931009-1G
APOSOR931009-1GEA
0
EUR
APOSOR931009-1G
4-(Boc-amino)pyridazine 95%
4-(Boc-amino)pyridazine
Formel:
C₉H₁₃N₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD16988479 CAS-Nummer: 169050-21-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...