6-Ethoxynicotinaldehyde 98%

Lieferant: Apollo Scientific

Synonyme: 2-Ethoxypyridine-5-carbaldehyde

OR931015-5G
APOSOR931015-5GEA 208 EUR
APOSOR931015-5G
6-Ethoxynicotinaldehyde 98%
6-Ethoxynicotinaldehyde
Formel: C₈H₉NO₂
Molecular Weight: 151.163 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11553587
CAS-Nummer: 97455-61-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR