[1-(2-Methyl-1,3-thiazol-4-yl)ethyl]amine dihydrochloride 95%

Lieferant: Apollo Scientific

Synonyme: 1-(2-Methyl-thiazol-4-yl)-ethylamine

OR931075-1G
APOSOR931075-1GEA 435 EUR
APOSOR931075-1G
[1-(2-Methyl-1,3-thiazol-4-yl)ethyl]amine dihydrochloride 95%
[1-(2-Methyl-1,3-thiazol-4-yl)ethyl]amine dihydrochloride
Formel: C₆H₁₂Cl₂N₂S
Molecular Weight: 215,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07781067
CAS-Nummer: 317830-81-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR