6-Methoxy-2-vinylnaphthalene 95%

Lieferant: Apollo Scientific

Synonyme: 2-Methoxy-6-vinylnaphthaleneyrol

OR931128-1G
APOSOR931128-1GEA 157 EUR
APOSOR931128-1G
6-Methoxy-2-vinylnaphthalene 95%
6-Methoxy-2-vinylnaphthalene
Formel: C₁₃H₁₂O
Molecular Weight: 184.234 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03427628
CAS-Nummer: 63444-51-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR