2-Chloro-1-(pyridin-3-yl)ethanone hydrochloride 95%
Lieferant: Apollo Scientific
OR931181-1G
APOSOR931181-1GEA
132
EUR
APOSOR931181-1G
2-Chloro-1-(pyridin-3-yl)ethanone hydrochloride 95%
2-Chloro-1-(pyridin-3-yl)ethanone hydrochloride
Formel:
C₇H₆ClNO·ClH Molecular Weight: 192.043 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09033889 CAS-Nummer: 61889-48-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...