Benzotriazol-1-ylacetic acid 96%

Lieferant: Apollo Scientific

Synonyme: 2-(1H-Benzo[d][1,2,3]triazol-1-yl)acetic acid

OR931189-1G
APOSOR931189-1GEA 209 EUR
APOSOR931189-1G
Benzotriazol-1-ylacetic acid 96%
Benzotriazol-1-ylacetic acid
Formel: C₈H₇N₃O₂
Molecular Weight: 177.16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00553342
CAS-Nummer: 4144-64-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR