1,4-Dimethoxy-2,3-dimethylbenzene 95%

Lieferant: Apollo Scientific

OR931246-1G
APOSOR931246-1GEA 140 EUR
APOSOR931246-1G
1,4-Dimethoxy-2,3-dimethylbenzene 95%
1,4-Dimethoxy-2,3-dimethylbenzene
Formel: C₁₀H₁₄O₂
Molecular Weight: 166,217 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00094721
CAS-Nummer: 39021-83-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR