3-(Cyanomethyl-amino)-benzoic acid methyl ester 95%

Lieferant: Apollo Scientific

OR931270-1G
APOSOR931270-1GEA 632 EUR
APOSOR931270-1G
3-(Cyanomethyl-amino)-benzoic acid methyl ester 95%
3-(Cyanomethyl-amino)-benzoic acid methyl ester
Formel: C₁₀H₁₀N₂O₂
Molecular Weight: 190,199 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16171448
CAS-Nummer: 75254-58-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR