5-Chloro-3-methyl-4-nitro-1H-pyrazole 95%

Lieferant: Apollo Scientific

OR931371-1G
APOSOR931371-1GEA 89.9 EUR
APOSOR931371-1G
5-Chloro-3-methyl-4-nitro-1H-pyrazole 95%
5-Chloro-3-methyl-4-nitro-1H-pyrazole
Formel: C₄H₄ClN₃O₂
Molecular Weight: 161.546 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00462202
CAS-Nummer: 6814-58-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR