3-(2-Nitrophenyl)propan-1-ol 95%

Lieferant: Apollo Scientific

OR931377-1G
APOSOR931377-1GEA 647 EUR
APOSOR931377-1G
3-(2-Nitrophenyl)propan-1-ol 95%
3-(2-Nitrophenyl)propan-1-ol
Formel: C₉H₁₁NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09028697
CAS-Nummer: 20716-26-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR