4-Isopropylcyclohexanecarboxylic acid (cis and trans mixture) 95%

Lieferant: Apollo Scientific

Synonyme: Isopropyl-cyclohexanecarboxylic acid

OR931378-5G
APOSOR931378-5GEA 342 EUR
APOSOR931378-5G
4-Isopropylcyclohexanecarboxylic acid (cis and trans mixture) 95%
4-Isopropylcyclohexanecarboxylic acid (cis and trans mixture)
Formel: C₁₀H₁₈O₂
Molecular Weight: 170,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01734696
CAS-Nummer: 62067-45-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR