3-Chloro-5-hydroxy-2-methoxypyridine 95%

Lieferant: Apollo Scientific

OR931387-1G OR931387-250MG
APOSOR931387-1GEA 828 EUR
APOSOR931387-1G APOSOR931387-250MG
3-Chloro-5-hydroxy-2-methoxypyridine 95%
3-Chloro-5-hydroxy-2-methoxypyridine
Formel: C₆H₆ClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD13190369
CAS-Nummer: 1196157-30-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR