6-Bromo-L-tryptophan 97%

Lieferant: Apollo Scientific

OR931390-1G OR931390-250MG
APOSOR931390-1GEA 0 EUR
APOSOR931390-1G APOSOR931390-250MG
6-Bromo-L-tryptophan 97%
6-Bromo-L-tryptophan
Formel: C₁₁H₁₁BrN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD17214433
CAS-Nummer: 52448-17-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR