(2-Amino-6-chlorophenyl)methanol 97%

Lieferant: Apollo Scientific
Achtung

OR931392-5G
APOSOR931392-5GEA 53.8 EUR
APOSOR931392-5G
(2-Amino-6-chlorophenyl)methanol 97%
(2-Amino-6-chlorophenyl)methanol
Formel: C₇H₈ClNO
Lagertemperatur: Raumtemperatur
MDL: MFCD09029932
CAS-Nummer: 39885-08-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR