Bis(trimethylsilyl)carbodiimid 95%
Lieferant: Apollo Scientific
OR931437-25G
OR931437-5G
APOSOR931437-25GEA
828
EUR
APOSOR931437-25G
APOSOR931437-5G
Bis(trimethylsilyl)carbodiimid 95%
Bis(trimethylsilyl)carbodiimid
Formel:
C₇H₁₈N₂Si₂ Molecular Weight: 186.4 g/mol Siedepunkt: 164 Schmelzpunkt: -23 Dichte: 0.8211 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051538 CAS-Nummer: 1000-70-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...