3-(2-Cyanopropan-2-yl)benzoic acid 95%
Lieferant: Apollo Scientific
OR931459-1G
OR931459-250MG
APOSOR931459-1GEA
87
EUR
APOSOR931459-1G
APOSOR931459-250MG
3-(2-Cyanopropan-2-yl)benzoic acid 95%
3-(2-Cyanopropan-2-yl)benzoic acid
Formel:
C₁₁H₁₁NO₂ Molecular Weight: 189.211 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD19694856 CAS-Nummer: 872091-00-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...