2-(4-Methylphenyl)ethanol 95%

Lieferant: Apollo Scientific

Synonyme: 2-(p-Tolyl)ethanol

OR931463-25G
APOSOR931463-25GEA 58 EUR
APOSOR931463-25G
2-(4-Methylphenyl)ethanol 95%
2-(4-Methylphenyl)ethanol
Formel: C₉H₁₂O
Molecular Weight: 136.20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00002904
CAS-Nummer: 699-02-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR