1-(3-Nitro-4-pyridinyl)-ethanone 95%
Lieferant: Apollo Scientific
Synonyme:
1-(3-Nitropyridin-4-yl)ethanone
OR931499-1G
APOSOR931499-1GEA
58
EUR
APOSOR931499-1G
1-(3-Nitro-4-pyridinyl)-ethanone 95%
1-(3-Nitro-4-pyridinyl)-ethanone
Formel:
C₇H₆N₂O₃ Molecular Weight: 166,134 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03411728 CAS-Nummer: 161871-65-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...