4-Chlor-3,5-xylenol 98%

Lieferant: Apollo Scientific

OR9315-25G OR9315-500G
APOSOR9315-25GEA 21.9 EUR
APOSOR9315-25G APOSOR9315-500G
4-Chlor-3,5-xylenol 98%
4-Chlor-3,5-xylenol
Formel: C₈H₉ClO
Schmelzpunkt: 114…116 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002324
CAS-Nummer: 88-04-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR