3-Bromo-2-methylpropan-1-ol 95%

Lieferant: Apollo Scientific

OR931511-1G
APOSOR931511-1GEA 403 EUR
APOSOR931511-1G
3-Bromo-2-methylpropan-1-ol 95%
3-Bromo-2-methylpropan-1-ol
Formel: C₄H₉BrO
Molecular Weight: 153,018 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01365869
CAS-Nummer: 40145-08-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR