tert-Butyl but-3-yn-1-ylcarbamate 95%

Lieferant: Apollo Scientific

OR931520-1G
APOSOR931520-1GEA 59.6 EUR
APOSOR931520-1G
tert-Butyl but-3-yn-1-ylcarbamate 95%
tert-Butyl but-3-yn-1-ylcarbamate
Formel: C₉H₁₅NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD10698150
CAS-Nummer: 149990-27-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR