1-Hydroxy-benzocyclobutene 95%

Lieferant: Apollo Scientific

Synonyme: Bicyclo[4.2.0]octa-1,3,5-trien-7-ol

OR931522-1G
APOSOR931522-1GEA 235 EUR
APOSOR931522-1G
1-Hydroxy-benzocyclobutene 95%
1-Hydroxy-benzocyclobutene
Formel: C₈H₈O
Molecular Weight: 120,149 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00798161
CAS-Nummer: 35447-99-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR