1H-Pyrazolo[3,4-b]pyridine-3-carbonitrile 95%

Lieferant: Apollo Scientific

OR931536-1G
APOSOR931536-1GEA 88.6 EUR
APOSOR931536-1G
1H-Pyrazolo[3,4-b]pyridine-3-carbonitrile 95%
1H-Pyrazolo[3,4-b]pyridine-3-carbonitrile
Formel: C₇H₄N₄
Lagertemperatur: Raumtemperatur
MDL: MFCD13176785
CAS-Nummer: 956010-88-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR