Diethyl-morpholin-2-ylmethyl-amine 95%

Lieferant: Apollo Scientific

OR931575-1G
APOSOR931575-1GEA 464 EUR
APOSOR931575-1G
Diethyl-morpholin-2-ylmethyl-amine 95%
Diethyl-morpholin-2-ylmethyl-amine
Formel: C₉H₂₀N₂O
Molecular Weight: 172,268 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03990578
CAS-Nummer: 122894-66-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR