4-Brom-N-methylanilin 98%

Lieferant: Apollo Scientific

OR931639-25G
APOSOR931639-25GEA 156 EUR
APOSOR931639-25G
4-Brom-N-methylanilin 98%
4-Brom-N-methylanilin
Formel: C₇H₈BrN
Molecular Weight: 186,05 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03030391
CAS-Nummer: 6911-87-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR