3-Formyl-1H-indole-6-carbonitrile ≥98%

Lieferant: Apollo Scientific

OR931695-5G
APOSOR931695-5GEA 137 EUR
APOSOR931695-5G
3-Formyl-1H-indole-6-carbonitrile ≥98%
3-Formyl-1H-indole-6-carbonitrile
Formel: C₁₀H₆N₂O
Molecular Weight: 170.167 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06657155
CAS-Nummer: 83783-33-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR