5-(4-Chloro-phenyl)-3,4-dihydro-2H-pyrrole 95%

Lieferant: Apollo Scientific

OR931706-1G
APOSOR931706-1GEA 464 EUR
APOSOR931706-1G
5-(4-Chloro-phenyl)-3,4-dihydro-2H-pyrrole 95%
5-(4-Chloro-phenyl)-3,4-dihydro-2H-pyrrole
Formel: C₁₀H₁₀ClN
Molecular Weight: 179,646 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02931038
CAS-Nummer: 22217-78-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR