Methyl 3-(3-pyridyl)propiolate 95%
Lieferant: Apollo Scientific
Synonyme:
Methyl 3-(3-Pyridyl)propiolate
OR931728-1G
APOSOR931728-1GEA
272
EUR
APOSOR931728-1G
Methyl 3-(3-pyridyl)propiolate 95%
Methyl 3-(3-pyridyl)propiolate
Formel:
C₉H₇NO₂ Molecular Weight: 161,157 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18205098 CAS-Nummer: 78584-30-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...