N,N-Diethyl-o-Toluidin 95%

Lieferant: Apollo Scientific

OR931742-25G OR931742-5G
APOSOR931742-25GEA 609 EUR
APOSOR931742-25G APOSOR931742-5G
N,N-Diethyl-o-Toluidin 95%
N,N-Diethyl-o-Toluidin
Formel: C₁₁H₁₇N
Lagertemperatur: Raumtemperatur
MDL: MFCD00059227
CAS-Nummer: 606-46-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR