1-(1H-Indol-2-yl)ethanone 95%
Lieferant: Apollo Scientific
Synonyme:
2-Acetylindole
OR931775-1G
APOSOR931775-1GEA
62.5
EUR
APOSOR931775-1G
1-(1H-Indol-2-yl)ethanone 95%
1-(1H-Indol-2-yl)ethanone
Formel:
C₁₀H₉NO Molecular Weight: 159,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01863329 CAS-Nummer: 4264-35-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...