Methyl 4-hydroxy-2,3-dimethylbenzoate 95%
Lieferant: Apollo Scientific
OR931778-1G
APOSOR931778-1GEA
719
EUR
APOSOR931778-1G
Methyl 4-hydroxy-2,3-dimethylbenzoate 95%
Methyl 4-hydroxy-2,3-dimethylbenzoate
Formel:
C₁₀H₁₂O₃ Molecular Weight: 180,2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13659355 CAS-Nummer: 5628-56-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...