4-Chloro-5-methyl-2-(methylthio)pyrimidine 95%

Lieferant: Apollo Scientific

OR931785-1G OR931785-250MG
APOSOR931785-1GEA 0 EUR
APOSOR931785-1G APOSOR931785-250MG
4-Chloro-5-methyl-2-(methylthio)pyrimidine 95%
4-Chloro-5-methyl-2-(methylthio)pyrimidine
Formel: C₆H₇ClN₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD09951961
CAS-Nummer: 61044-96-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR