Methyl-4-(hydroxymethyl)picolinat 95%
Lieferant: Apollo Scientific
OR931790-1G
APOSOR931790-1GEA
120
EUR
APOSOR931790-1G
Methyl-4-(hydroxymethyl)picolinat 95%
Methyl-4-(hydroxymethyl)picolinat
Formel:
C₈H₉NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09834350 CAS-Nummer: 317335-15-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...