(5-Chloro-pyrimidin-2-yl)-cyclopropyl-amine 95%

Lieferant: Apollo Scientific

OR931825-1G
APOSOR931825-1GEA 0 EUR
APOSOR931825-1G
(5-Chloro-pyrimidin-2-yl)-cyclopropyl-amine 95%
(5-Chloro-pyrimidin-2-yl)-cyclopropyl-amine
Formel: C₇H₈ClN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD18837178
CAS-Nummer: 1289385-19-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR