(2-Chloro-6-methyl-pyrimidin-4-yl)-cyclopropyl-amine 95%
Lieferant: Apollo Scientific
OR931917-1G
APOSOR931917-1GEA
0
EUR
APOSOR931917-1G
(2-Chloro-6-methyl-pyrimidin-4-yl)-cyclopropyl-amine 95%
(2-Chloro-6-methyl-pyrimidin-4-yl)-cyclopropyl-amine
Formel:
C₈H₁₀ClN₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD16767268 CAS-Nummer: 1250038-98-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...