α,α,2,4-tetrachlorotoluene 98%

Lieferant: Apollo Scientific

Synonyme: 2,4-Dichloro-1-(dichloromethyl)benzene

OR931922-5G
APOSOR931922-5GEA 68.3 EUR
APOSOR931922-5G
α,α,2,4-tetrachlorotoluene 98%
α,α,2,4-tetrachlorotoluene
Formel: C₇H₄Cl₄
Molecular Weight: 229.919 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00043967
CAS-Nummer: 134-25-8
UN: 3077
ADR: 9+EHS III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR